siRNA Details
| Gene Symbol: DHRS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144683.3 | NP_653284.2 | 5 | 1199 | ||
| GenBank mRNA | AK296017.1 | - | 3 | 850 | ||
| AY358566.1 | - | 5 | 1199 | |||
| BC015582.2 | - | 4 | 1189 | 4637504 | ||
| BQ052759.1 | - | 1 | 19 | |||

DHRS13
Genomic Map