siRNA Details
| Gene Symbol: DHRSX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145177.2 | NP_660160.2 | 3 | 295 | ||
| GenBank mRNA | AJ293620.1 | - | 3 | 255 | ||
| AY358849.1 | - | 3 | 257 | |||
| BC019696.2 | - | 3 | 248 | |||
| BC032340.1 | - | 3 | 259 | 5192975 | ||

DHRSX
Genomic Map