siRNA Details
| Gene Symbol: DHRSX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145177.2 | NP_660160.2 | 6 | 837 | ||
| GenBank mRNA | AJ293620.1 | - | 6 | 797 | ||
| AL137300.1 | - | 2 | 411 | |||
| AY358849.1 | - | 6 | 799 | |||
| BC019696.2 | - | 6 | 790 | |||
| BC032340.1 | - | 6 | 801 | 5192975 | ||
| BM794858.1 | - | 3 | 517 | |||

DHRSX
Genomic Map