siRNA Details
| Gene Symbol: SENP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308045.1 | NP_001294974.1 | 6 | 2080 | ||
| NM_152699.4 | NP_689912.2 | 6 | 2080 | |||
| XM_005269310.2 | XP_005269367.1 | 6 | 2080 | |||
| XM_005269312.2 | XP_005269369.1 | 6 | 2080 | |||
| XM_011512544.2 | XP_011510846.1 | 7 | 2125 | |||
| XM_011512545.2 | XP_011510847.1 | 6 | 2080 | |||
| XM_017005870.1 | XP_016861359.1 | 7 | 2080 | |||
| GenBank mRNA | AF335474.1 | - | 5 | 937 | ||
| AK124389.1 | - | 5 | 1053 | |||
| AK302305.1 | - | 6 | 2080 | |||
| AK313764.1 | - | 6 | 2053 | |||
| BC000958.2 | - | 5 | 1177 | |||
| BC008589.1 | - | 5 | 1177 | |||
| BC030705.2 | - | 6 | 2029 | 4824344 | ||

SENP5
Genomic Map