siRNA Details
| Gene Symbol: HS3ST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153612.3 | NP_705840.2 | 2 | 442 | ||
| XM_006715379.2 | XP_006715442.1 | 5 | 930 | |||
| XM_011535588.2 | XP_011533890.1 | 3 | 1646 | |||
| XM_017010470.1 | XP_016865959.1 | 5 | 1504 | |||
| XM_017010471.1 | XP_016865960.1 | 5 | 2463 | |||
| XM_017010472.1 | XP_016865961.1 | 4 | 2100 | |||
| XM_017010473.1 | XP_016865962.1 | 5 | 1500 | |||
| XM_017010474.1 | XP_016865963.1 | 5 | 938 | |||
| GenBank mRNA | AF503292.1 | - | 2 | 410 | ||
| AK126930.1 | - | 2 | 1102 | |||
| AK289907.1 | - | 5 | 929 | |||
| BC093911.1 | - | 2 | 443 | 7939756 | ||
| BC093913.1 | - | 2 | 443 | 7939758 | ||

HS3ST5
Genomic Map