siRNA Details
| Gene Symbol: CACNA2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172364.4 | NP_758952.4 | 2 | 517 | ||
| XM_011521041.2 | XP_011519343.1 | 2 | 4419 | |||
| GenBank mRNA | AF516695.1 | - | 2 | 649 | ||
| BC150186.1 | - | 2 | 450 | 40148321 | ||
| BX537437.1 | - | 2 | 518 | |||

CACNA2D4
Genomic Map