siRNA Details
| Gene Symbol: HSD17B13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178135.4 | NP_835236.2 | 2 | 305 | ||
| GenBank mRNA | AB073347.1 | - | 2 | 274 | ||
| AK292784.1 | - | 2 | 279 | |||
| AY186249.1 | - | 2 | 285 | |||
| AY268355.1 | - | 2 | 255 | |||
| AY358575.1 | - | 2 | 305 | |||
| BC112303.1 | - | 2 | 264 | 8327771 | ||
| BC112305.1 | - | 2 | 264 | 8327773 | ||
| DR004209.1 | - | 2 | 310 | |||

HSD17B13
Genomic Map