siRNA Details
| Gene Symbol: PKD1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181536.1 | NP_853514.1 | 30 | 5102 | ||
| GenBank mRNA | AY164485.1 | - | 30 | 5102 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKD1L3| Gene Symbol: PKD1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181536.1 | NP_853514.1 | 30 | 5102 | ||
| GenBank mRNA | AY164485.1 | - | 30 | 5102 | ||