siRNA Details
Gene Symbol: KCNK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181840.1 | NP_862823.1 | 2 | 322 | ||
GenBank mRNA | AB087138.1 | - | 2 | 322 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KCNK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181840.1 | NP_862823.1 | 2 | 322 | ||
GenBank mRNA | AB087138.1 | - | 2 | 322 |