siRNA Details
| Gene Symbol: TMPRSS11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114387.1 | NP_001107859.1 | 2 | 230 | ||
| NM_182606.3 | NP_872412.3 | 2 | 230 | |||
| GenBank mRNA | AF071882.1 | - | 2 | 190 | ||
| AK131518.1 | - | 2 | 230 | |||
| AK292950.1 | - | 2 | 168 | |||
| AY498712.1 | - | 2 | 142 | |||
| BC111796.1 | - | 2 | 156 | 40026103 | ||

TMPRSS11A
Genomic Map