siRNA Details
| Gene Symbol: TMPRSS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182973.1 | NP_892018.1 | 15 | 2720 | ||
| XM_011527978.2 | XP_011526280.1 | 17 | 3083 | |||
| XM_011527980.1 | XP_011526282.1 | 9 | 1739 | |||
| XM_011527982.2 | XP_011526284.1 | 7 | 1514 | |||
| GenBank mRNA | AJ488946.1 | - | 15 | 2720 | ||
| AK131261.1 | - | 7 | 1308 | |||

TMPRSS9
Genomic Map