siRNA Details
| Gene Symbol: POU3F4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000307.4 | NP_000298.3 | 1 | 1098 | ||
| GenBank mRNA | AK314967.1 | - | 1 | 1098 | ||
| X82324.1 | - | 1 | 1066 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU3F4| Gene Symbol: POU3F4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000307.4 | NP_000298.3 | 1 | 1098 | ||
| GenBank mRNA | AK314967.1 | - | 1 | 1098 | ||
| X82324.1 | - | 1 | 1066 | |||