siRNA Details
| Gene Symbol: MMP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002428.3 | NP_002419.1 | 3 | 1203 | ||
| GenBank mRNA | BC006412.1 | - | 3 | 323 | ||
| BC036495.1 | - | 3 | 1209 | 5263145 | ||
| BC055428.1 | - | 3 | 1204 | 4811572 | ||
| BT006715.1 | - | 1 | 103 | |||
| D86331.1 | - | 2 | 141 | |||
| Z48482.1 | - | 3 | 466 | |||

MMP15
Genomic Map