siRNA Details
| Gene Symbol: MN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002430.2 | NP_002421.3 | 1 | 1723 | ||
| GenBank mRNA | X82209.2 | - | 1 | 1724 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MN1| Gene Symbol: MN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002430.2 | NP_002421.3 | 1 | 1723 | ||
| GenBank mRNA | X82209.2 | - | 1 | 1724 | ||