siRNA Details
| Gene Symbol: EN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001426.3 | NP_001417.3 | 2 | 2105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EN1| Gene Symbol: EN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001426.3 | NP_001417.3 | 2 | 2105 | ||