siRNA Details
| Gene Symbol: DUSP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001947.3 | NP_001938.2 | 3 | 1314 | ||
| GenBank mRNA | AF508727.1 | - | 3 | 1082 | ||
| AF508728.1 | - | 1 | 530 | |||
| BC019107.2 | - | 3 | 946 | |||
| BC104880.1 | - | 3 | 1142 | 8143883 | ||
| BC104882.1 | - | 3 | 1142 | 8143885 | ||
| X93921.1 | - | 3 | 944 | |||

DUSP7
Genomic Map