siRNA Details
| Gene Symbol: HOXD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002148.3 | NP_002139.2 | 2 | 1188 | ||
| GenBank mRNA | BX114645.1 | - | 2 | 556 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXD10| Gene Symbol: HOXD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002148.3 | NP_002139.2 | 2 | 1188 | ||
| GenBank mRNA | BX114645.1 | - | 2 | 556 | ||