siRNA Details
| Gene Symbol: PBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204961.1 | NP_001191890.1 | 8 | 1762 | ||
| NM_002585.3 | NP_002576.1 | 9 | 1875 | |||
| XM_005245228.3 | XP_005245285.1 | 10 | 2043 | |||
| XM_005245229.3 | XP_005245286.1 | 10 | 1636 | |||
| XM_011509591.2 | XP_011507893.1 | 9 | 1930 | |||
| GenBank mRNA | AK291415.1 | - | 9 | 1557 | ||
| BX647313.1 | - | 3 | 2470 | |||
| CR749446.1 | - | 3 | 7760 | |||
| M31170.1 | - | 7 | 1148 | |||
| M86546.1 | - | 9 | 1534 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001291509, NM_001291508, NM_183355, L27453, AF020196, and others.

PBX1
Genomic Map