siRNA Details
| Gene Symbol: PTPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145369.1 | NP_001138841.1 | 7, 8 | 3164 | ||
| NM_001145371.1 | NP_001138843.1 | 2, 3 | 598 | |||
| NM_002829.3 | NP_002820.3 | 12, 13 | 1115 | |||
| XM_006717197.3 | XP_006717260.1 | 12, 13 | 1224 | |||
| XM_006717199.3 | XP_006717262.1 | 12, 13 | 1249 | |||
| XM_006717201.3 | XP_006717264.1 | 11, 12 | 1056 | |||
| XM_006717202.3 | XP_006717265.1 | 9, 10 | 818 | |||
| XM_006717203.3 | XP_006717266.1 | 9, 10 | 777 | |||
| XM_006717204.3 | XP_006717267.1 | 12, 13 | 1151 | |||
| XM_011518888.2 | XP_011517190.1 | 12, 13 | 1185 | |||
| XM_011518889.2 | XP_011517191.1 | 12, 13 | 1855 | |||
| XM_017014955.1 | XP_016870444.1 | 12, 13 | 1207 | |||
| GenBank mRNA | AK296388.1 | - | 2, 3 | 524 | ||
| BC126117.1 | - | 11, 12 | 1025 | 8991833 | ||
| BX648735.1 | - | 7, 8 | 1599 | |||
| DQ104439.1 | - | 11, 12 | 1003 | |||
| M64572.1 | - | 11, 12 | 1026 | |||
| S39392.1 | - | 4, 5 | 426 | |||

PTPN3
Genomic Map