siRNA Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003083.3 | NP_003074.1 | 5 | 771 | ||
| NR_030717.1 | 5 | 668 | ||||
| GenBank mRNA | AK222926.1 | - | 5 | 742 | ||
| AK314878.1 | - | 5 | 756 | |||
| BC011868.2 | - | 5 | 726 | 4545178 | ||
| BX647416.1 | - | 5 | 635 | |||
| U44755.1 | - | 5 | 755 | |||
| U44898.1 | - | 5 | 737 | |||

SNAPC2
Genomic Map