siRNA Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003083.3 | NP_003074.1 | 5 | 966 | ||
| NR_030717.1 | 5 | 863 | ||||
| GenBank mRNA | AK222926.1 | - | 5 | 937 | ||
| AK314878.1 | - | 5 | 951 | |||
| BC011868.2 | - | 5 | 921 | 4545178 | ||
| BX647416.1 | - | 5 | 830 | |||
| U44755.1 | - | 5 | 950 | |||
| U44898.1 | - | 5 | 932 | |||

SNAPC2
Genomic Map