siRNA Details
| Gene Symbol: PHOX2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003924.3 | NP_003915.2 | 3 | 1904 | ||
| GenBank mRNA | D82344.1 | - | 3 | 1904 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHOX2B| Gene Symbol: PHOX2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003924.3 | NP_003915.2 | 3 | 1904 | ||
| GenBank mRNA | D82344.1 | - | 3 | 1904 | ||