siRNA Details
| Gene Symbol: RAB3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004283.3 | NP_004274.1 | 5 | 1441 | ||
| GenBank mRNA | AF019226.1 | - | 1 | 469 | ||
| BC016471.1 | - | 5 | 1418 | 3861912 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB3D| Gene Symbol: RAB3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004283.3 | NP_004274.1 | 5 | 1441 | ||
| GenBank mRNA | AF019226.1 | - | 1 | 469 | ||
| BC016471.1 | - | 5 | 1418 | 3861912 | ||