siRNA Details
| Gene Symbol: ELF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114309.1 | NP_001107781.1 | 4 | 680 | ||
| NM_004433.4 | NP_004424.3 | 4 | 566 | |||
| XM_005244942.2 | XP_005244999.1 | 4 | 609 | |||
| GenBank mRNA | AF016295.1 | - | 4 | 527 | ||
| AF017307.1 | - | 4 | 615 | |||
| AF517841.1 | - | 4 | 548 | |||
| AK225446.1 | - | 4 | 503 | |||
| AK309265.1 | - | 2 | 539 | |||
| AK310814.1 | - | 4 | 558 | |||
| AK312273.1 | - | 4 | 542 | |||
| BC003569.1 | - | 4 | 534 | 3537846 | ||
| BX537368.1 | - | 3 | 3603 | |||
| CR457106.1 | - | 3 | 413 | |||
| CV570991.1 | - | 4 | 566 | |||
| U66894.1 | - | 4 | 508 | |||
| U73843.1 | - | 4 | 532 | |||
| U73844.1 | - | 4 | 532 | |||
| U97156.1 | - | 4 | 537 | |||

ELF3
Genomic Map