siRNA Details
| Gene Symbol: ELF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114309.1 | NP_001107781.1 | 9 | 1855 | ||
| NM_004433.4 | NP_004424.3 | 9 | 1741 | |||
| GenBank mRNA | AF016295.1 | - | 9 | 1702 | ||
| AF017307.1 | - | 9 | 1787 | |||
| AF517841.1 | - | 9 | 1726 | |||
| BC003569.1 | - | 9 | 1709 | 3537846 | ||
| BX537368.1 | - | 8 | 4778 | |||
| U66894.1 | - | 9 | 1681 | |||
| U73843.1 | - | 9 | 1707 | |||
| U73844.1 | - | 10 | 1638 | |||

ELF3
Genomic Map