siRNA Details
| Gene Symbol: PITX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005029.3 | NP_005020.1 | 3 | 327 | ||
| GenBank mRNA | AF041339.1 | - | 2 | 176 | ||
| BC011642.2 | - | 3 | 326 | 4131442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PITX3| Gene Symbol: PITX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005029.3 | NP_005020.1 | 3 | 327 | ||
| GenBank mRNA | AF041339.1 | - | 2 | 176 | ||
| BC011642.2 | - | 3 | 326 | 4131442 | ||