siRNA Details
| Gene Symbol: PHOX2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005169.3 | NP_005160.2 | 3 | 1238 | ||
| GenBank mRNA | AK290645.1 | - | 3 | 1238 | ||
| BC041564.1 | - | 3 | 1220 | 5923445 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHOX2A| Gene Symbol: PHOX2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005169.3 | NP_005160.2 | 3 | 1238 | ||
| GenBank mRNA | AK290645.1 | - | 3 | 1238 | ||
| BC041564.1 | - | 3 | 1220 | 5923445 | ||