siRNA Details
Gene Symbol: CDKN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005192.3 | NP_005183.2 | 3 | 228 | ||
XM_011536353.2 | XP_011534655.1 | 3 | 244 | |||
GenBank mRNA | AF213033.1 | - | 2 | 114 | ||
AF213035.1 | - | 2 | 114 | |||
AF213036.1 | - | 2 | 114 | |||
AF213037.1 | - | 2 | 114 | |||
AF213039.1 | - | 2 | 114 | |||
AF213040.1 | - | 2 | 114 | |||
AF213041.1 | - | 2 | 114 | |||
AF213046.1 | - | 2 | 114 | |||
AF213050.1 | - | 2 | 114 | |||
AF213052.1 | - | 2 | 114 | |||
AF213053.1 | - | 2 | 114 | |||
AK098653.1 | - | 3 | 228 | |||
AK293265.1 | - | 3 | 154 | |||
AY257474.1 | - | 2 | 114 | |||
AY257475.1 | - | 2 | 114 | |||
AY257476.1 | - | 2 | 114 | |||
AY257478.1 | - | 2 | 114 | |||
BC103633.1 | - | 3 | 160 | |||
BT019451.1 | - | 2 | 114 | |||
CR407666.1 | - | 2 | 114 | |||
EF560750.1 | - | 3 | 154 | |||
KP966095.1 | - | 2 | 145 | |||
L25876.1 | - | 3 | 175 | |||
L27711.1 | - | 3 | 165 | |||
U02681.1 | - | 3 | 145 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
