siRNA Details
| Gene Symbol: CDX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005193.1 | NP_005184.1 | 1 | 150 | ||
| GenBank mRNA | BC128233.1 | - | 1 | 150 | 40114658 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDX4| Gene Symbol: CDX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005193.1 | NP_005184.1 | 1 | 150 | ||
| GenBank mRNA | BC128233.1 | - | 1 | 150 | 40114658 | |