siRNA Details
| Gene Symbol: CHRNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005199.4 | NP_005190.4 | 6 | 595 | ||
| GenBank mRNA | AK125362.1 | - | 6 | 595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRNG| Gene Symbol: CHRNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005199.4 | NP_005190.4 | 6 | 595 | ||
| GenBank mRNA | AK125362.1 | - | 6 | 595 | ||