siRNA Details
| Gene Symbol: TACSTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002353.2 | NP_002344.2 | 1 | 1270 | ||
| GenBank mRNA | BC009409.1 | - | 1 | 1017 | 3611756 | |
| BM765888.1 | - | 1 | 373 | |||
| BT007255.1 | - | 1 | 932 | |||
| CR542260.1 | - | 1 | 932 | |||
| X13425.1 | - | 1 | 1002 | |||
| X77753.1 | - | 1 | 1547 | |||

TACSTD2
Genomic Map