siRNA Details
| Gene Symbol: INPP5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321042.1 | NP_001307971.1 | 13 | 2693 | ||
| NM_005539.4 | NP_005530.3 | 16 | 2640 | |||
| XM_017016204.1 | XP_016871693.1 | 15 | 2294 | |||
| GenBank mRNA | BC052265.1 | - | 13 | 2066 | ||
| BC062300.1 | - | 13 | 2066 | |||
| BI964266.1 | - | 1 | 18 | |||
| Z31695.1 | - | 16 | 2273 | |||

INPP5A
Genomic Map