siRNA Details
| Gene Symbol: SPRY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258038.1 | NP_001244967.1 | 3 | 2559 | ||
| NM_001258039.1 | NP_001244968.1 | 2 | 2303 | |||
| NM_005841.2 | NP_005832.1 | 2 | 2480 | |||
| NM_199327.2 | NP_955359.1 | 2 | 2313 | |||
| XM_005262686.1 | XP_005262743.1 | 3 | 3220 | |||
| XM_017007635.1 | XP_016863124.1 | 2 | 2293 | |||
| GenBank mRNA | AF041037.1 | - | 1 | 1545 | ||
| AK026960.1 | - | 2 | 2457 | |||
| AY192146.1 | - | 2 | 2308 | |||
| AY590694.1 | - | 1 | 2111 | |||
| BC063856.1 | - | 2 | 2442 | 6181674 | ||

SPRY1
Genomic Map