siRNA Details
| Gene Symbol: PAX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006193.2 | NP_006184.2 | 4 | 703 | ||
| XM_011516276.2 | XP_011514578.1 | 7 | 977 | |||
| GenBank mRNA | AB008913.1 | - | 4 | 521 | ||
| AF043978.1 | - | 4 | 703 | |||
| BC074761.2 | - | 4 | 530 | 30915220 | ||
| BC107149.2 | - | 3 | 952 | 40032097 | ||
| BC107150.1 | - | 3 | 952 | 40032102 | ||
| DQ442656.1 | - | 3 | 491 | |||

PAX4
Genomic Map