siRNA Details
| Gene Symbol: MAGOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002370.3 | NP_002361.1 | 3 | 344 | ||
| GenBank mRNA | AF035940.1 | - | 3 | 247 | ||
| AF067173.1 | - | 3 | 270 | |||
| AK312113.1 | - | 3 | 280 | |||
| BC018211.1 | - | 3 | 256 | 3861094 | ||
| BX283982.1 | - | 3 | 344 | |||

MAGOH
Genomic Map