siRNA Details
| Gene Symbol: MAN2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002372.3 | NP_002363.2 | 2 | 1349 | ||
| XM_011543395.2 | XP_011541697.1 | 2 | 788 | |||
| XM_017009472.1 | XP_016864961.1 | 2 | 270 | |||
| GenBank mRNA | AK314090.1 | - | 2 | 690 | ||
| BC142656.1 | - | 2 | 651 | 40148064 | ||
| BC142696.1 | - | 2 | 651 | 40148740 | ||
| D63998.1 | - | 2 | 807 | |||
| U31520.1 | - | 2 | 300 | |||

MAN2A1
Genomic Map