siRNA Details
| Gene Symbol: PPP2R5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199756.1 | NP_001186685.1 | 2 | 469 | ||
| NM_006243.3 | NP_006234.1 | 2 | 937 | |||
| GenBank mRNA | AK302202.1 | - | 2 | 469 | ||
| AK312530.1 | - | 3 | 752 | |||
| BC022474.1 | - | 2 | 523 | 4815091 | ||
| BC110883.1 | - | 2 | 646 | 6045358 | ||
| BP326450.1 | - | 2 | 462 | |||
| L42373.1 | - | 2 | 934 | |||

PPP2R5A
Genomic Map