siRNA Details
| Gene Symbol: ARID3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307939.1 | NP_001294868.1 | 2 | 645 | ||
| NM_006465.3 | NP_006456.1 | 2 | 645 | |||
| GenBank mRNA | AB208830.1 | - | 2 | 564 | ||
| AF116846.1 | - | 2 | 577 | |||
| AK298716.1 | - | 2 | 570 | |||
| AK302034.1 | - | 2 | 645 | |||
| BC041792.1 | - | 2 | 573 | 5297774 | ||
| BC045545.1 | - | 2 | 619 | |||
| BC060824.1 | - | 2 | 558 | |||

ARID3B
Genomic Map