siRNA Details
| Gene Symbol: COPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006837.2 | NP_006828.2 | 2 | 486 | ||
| GenBank mRNA | AA203444.1 | - | 3 | 403 | ||
| AB209134.1 | - | 1 | 2196 | |||
| AK056967.1 | - | 4 | 262 | |||
| AK128519.1 | - | 1 | 2478 | |||
| AK310434.1 | - | 2 | 288 | |||
| AK311071.1 | - | 2 | 290 | |||
| BC001187.1 | - | 2 | 290 | 3354130 | ||
| BC001859.2 | - | 2 | 268 | 3530890 | ||
| BC007272.1 | - | 2 | 264 | 3161025 | ||
| BI752436.1 | - | 476 | ||||
| BX648542.1 | - | 2 | 2823 | |||
| CR541678.1 | - | 2 | 155 | |||
| U65928.1 | - | 2 | 282 | |||
| U70734.1 | - | 2 | 275 | |||

COPS5
Genomic Map