siRNA Details
| Gene Symbol: GTF3C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206774.1 | NP_001193703.1 | 4 | 631 | ||
| NM_012086.4 | NP_036218.1 | 4 | 631 | |||
| XM_005246965.4 | XP_005247022.1 | 4 | 546 | |||
| GenBank mRNA | AF133123.1 | - | 4 | 567 | ||
| AF465407.1 | - | 4 | 522 | |||
| AK025560.1 | - | 4 | 545 | |||
| AK314718.1 | - | 4 | 572 | |||
| BC043347.1 | - | 4 | 486 | 5498609 | ||

GTF3C3
Genomic Map