siRNA Details
| Gene Symbol: RBPJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281448.1 | NP_001268377.1 | 6 | 552 | ||
| NM_001281449.1 | NP_001268378.1 | 6 | 552 | |||
| NM_014276.3 | NP_055091.2 | 6 | 552 | |||
| XM_011528521.2 | XP_011526823.1 | 6 | 559 | |||
| XM_011528522.2 | XP_011526824.1 | 6 | 559 | |||
| XM_011528523.2 | XP_011526825.1 | 6 | 559 | |||
| GenBank mRNA | AB024964.1 | - | 6 | 472 | ||
| AB026048.1 | - | 6 | 472 | |||
| AB027710.1 | - | 6 | 472 | |||
| BM311999.1 | - | 6 | 555 | |||
| CA950939.1 | - | 6 | 551 | |||

RBPJL
Genomic Map