siRNA Details
| Gene Symbol: MFAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135247.1 | NP_001128719.1 | 8 | 521 | ||
| NM_001135248.1 | NP_001128720.1 | 8 | 540 | |||
| NM_002403.3 | NP_002394.1 | 8 | 543 | |||
| NM_017459.2 | NP_059453.1 | 8 | 524 | |||
| XM_011541476.2 | XP_011539778.1 | 8 | 1150 | |||
| GenBank mRNA | AK222751.1 | - | 8 | 502 | ||
| BC015039.1 | - | 8 | 543 | 3910064 | ||
| CR407678.1 | - | 7 | 410 | |||
| DN996554.1 | - | 8 | 493 | |||
| U19718.1 | - | 7 | 451 | |||

MFAP2
Genomic Map