siRNA Details
| Gene Symbol: TAF9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015975.4 | NP_057059.2 | 5 | 542 | ||
| GenBank mRNA | AF220509.1 | - | 5 | 493 | ||
| AK312579.1 | - | 5 | 501 | |||
| BC009566.1 | - | 5 | 493 | 3996687 | ||
| BC010350.1 | - | 5 | 507 | 4276335 | ||
| BC071649.1 | - | 5 | 479 | 5246715 | ||
| BC146952.1 | - | 5 | 523 | 9056880 | ||
| BC146960.1 | - | 5 | 523 | 9056888 | ||
| BI544376.1 | - | 5 | 571 | |||

TAF9B
Genomic Map