siRNA Details
| Gene Symbol: PER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289861.1 | NP_001276790.1 | 3, 4 | 546 | ||
| NM_001289862.1 | NP_001276791.1 | 3, 4 | 546 | |||
| NM_001289863.1 | NP_001276792.1 | 3, 4 | 546 | |||
| NM_001289864.1 | NP_001276793.1 | 3, 4 | 546 | |||
| NM_016831.2 | NP_058515.1 | 3, 4 | 546 | |||
| XM_005263524.3 | XP_005263581.1 | 2, 3 | 421 | |||
| XM_011542385.2 | XP_011540687.1 | 2, 3 | 421 | |||
| XM_011542390.2 | XP_011540692.1 | 2, 3 | 421 | |||
| XM_017002723.1 | XP_016858212.1 | 2, 3 | 421 | |||
| XM_017002724.1 | XP_016858213.1 | 2, 3 | 421 | |||
| XM_017002725.1 | XP_016858214.1 | 2, 3 | 421 | |||
| XM_017002726.1 | XP_016858215.1 | 2, 3 | 421 | |||
| XM_017002727.1 | XP_016858216.1 | 4, 5 | 740 | |||
| XM_017002733.1 | XP_016858222.1 | 3, 4 | 598 | |||
| XM_017002734.1 | XP_016858223.1 | 4, 5 | 740 | |||
| XM_017002735.1 | XP_016858224.1 | 3, 4 | 598 | |||
| XM_017002736.1 | XP_016858225.1 | 4, 5 | 740 | |||
| GenBank mRNA | AB047686.1 | - | 2, 3 | 506 | ||
| AK307760.1 | - | 2, 3 | 421 | |||
| AK308303.1 | - | 3, 4 | 595 | |||
| AL157954.1 | - | 2, 3 | 457 | |||
| AY493418.1 | - | 2, 3 | 353 | |||
| BC026102.1 | - | 3, 4 | 568 | |||
| BC144137.1 | - | 2, 3 | 450 | |||
| BC146781.1 | - | 2, 3 | 506 | 9007235 | ||

PER3
Genomic Map