siRNA Details
| Gene Symbol: HOXC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017409.3 | NP_059105.2 | 1 | 189 | ||
| GenBank mRNA | AF255675.1 | - | 1 | 91 | ||
| AK313759.1 | - | 1 | 205 | |||
| AL558161.3 | - | 1 | 189 | |||
| BC001293.1 | - | 1 | 162 | 3458115 | ||
| BT009921.1 | - | 1 | 91 | |||

HOXC10
Genomic Map