siRNA Details
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017677.3 | NP_060147.2 | 2 | 168 | ||
GenBank mRNA | BC012399.1 | - | 2 | 146 | 3864128 | |
DA691850.1 | - | 2 | 168 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017677.3 | NP_060147.2 | 2 | 168 | ||
GenBank mRNA | BC012399.1 | - | 2 | 146 | 3864128 | |
DA691850.1 | - | 2 | 168 |