siRNA Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019104.2 | NP_061977.1 | 3 | 502 | ||
| GenBank mRNA | AK293116.1 | - | 3 | 267 | ||
| AK301992.1 | - | 3 | 502 | |||
| BC009071.1 | - | 3 | 221 | 3872616 | ||
| BI767008.1 | - | 3 | 252 | |||
| CB152701.1 | - | 3 | 304 | |||
| DC396639.1 | - | 3 | 502 | |||

LIN37
Genomic Map