siRNA Details
| Gene Symbol: PGLYRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020393.3 | NP_065126.2 | 3 | 444 | ||
| XM_011509789.2 | XP_011508091.1 | 3 | 456 | |||
| XM_011509790.1 | XP_011508092.1 | 3 | 444 | |||
| XM_011509791.2 | XP_011508093.1 | 3 | 447 | |||
| XM_011509792.1 | XP_011508094.1 | 3 | 444 | |||
| GenBank mRNA | AF242518.1 | - | 3 | 172 | ||
| AK292203.1 | - | 3 | 447 | |||
| AY035377.1 | - | 3 | 149 | |||
| BC107157.1 | - | 2 | 131 | 40032748 | ||
| BC107158.1 | - | 2 | 131 | 40032750 | ||
| BC142636.1 | - | 3 | 419 | 40147772 | ||

PGLYRP4
Genomic Map