siRNA Details
| Gene Symbol: LHPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167880.1 | NP_001161352.1 | 6 | 1378 | ||
| NM_001318331.1 | NP_001305260.1 | 4 | 1221 | |||
| NM_022126.3 | NP_071409.3 | 7 | 1470 | |||
| XM_006717943.1 | XP_006718006.1 | 8 | 1491 | |||
| GenBank mRNA | AK055532.1 | - | 7 | 1399 | ||
| BC110344.1 | - | 6 | 1332 | 5762731 | ||
| BC113629.1 | - | 8 | 1408 | 8322681 | ||
| BC113631.1 | - | 8 | 1391 | 8322683 | ||
| EF151005.1 | - | 2 | 978 | |||

LHPP
Genomic Map