siRNA Details
| Gene Symbol: HOXC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022658.3 | NP_073149.1 | 2 | 1495 | ||
| GenBank mRNA | BC053898.1 | - | 2 | 1496 | 6171090 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXC8| Gene Symbol: HOXC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022658.3 | NP_073149.1 | 2 | 1495 | ||
| GenBank mRNA | BC053898.1 | - | 2 | 1496 | 6171090 | |